Anti-SHANK1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309857EA 636 EUR
ORIGTA309857
Anti-SHANK1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: SHANK1 (SH3 and multiple ankyrin repeat domains 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR