Anti-PTPN5 Mouse Monoclonal Antibody [clone: 23E5]

Lieferant: OriGene
ORIGTA309823EA 668 EUR
ORIGTA309823
Anti-PTPN5 Mouse Monoclonal Antibody [clone: 23E5]
Antikörper
Type: Primary
Antigen: PTPN5 (protein tyrosine phosphatase, non-receptor type 5 (striatum-enriched))
Clonality: Monoclonal
Clone: 23E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR