Anti-PRDM1 Mouse Monoclonal Antibody [clone: 3H2-E8]

Lieferant: OriGene
ORIGTA309819EA 723 EUR
ORIGTA309819
Anti-PRDM1 Mouse Monoclonal Antibody [clone: 3H2-E8]
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: PRDM1 (PR domain containing 1, with ZNF domain)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR