Anti-PMP22 Mouse Monoclonal Antibody [clone: Hu1]

Lieferant: OriGene
ORIGTA309809EA 636 EUR
ORIGTA309809
Anti-PMP22 Mouse Monoclonal Antibody [clone: Hu1]
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: PMP22 (peripheral myelin protein 22)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR