Anti-PHF2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309800EA 544 EUR
ORIGTA309800
Anti-PHF2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: PHF2 (PHD finger protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR