Anti-NPFFR1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309771EA 544 EUR
ORIGTA309771
Anti-NPFFR1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: NPFFR1 (neuropeptide FF receptor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR