Anti-MGMT Mouse Monoclonal Antibody [clone: MT 23.2]

Lieferant: OriGene
ORIGTA309748EA 668 EUR
ORIGTA309748
Anti-MGMT Mouse Monoclonal Antibody [clone: MT 23.2]
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: MGMT (O-6-methylguanine-DNA methyltransferase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR