Anti-MFN1 Mouse Monoclonal Antibody [clone: 11E9-1H12]

Lieferant: OriGene
ORIGTA309747EA 668 EUR
ORIGTA309747
Anti-MFN1 Mouse Monoclonal Antibody [clone: 11E9-1H12]
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: MFN1 (mitofusin 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR