Anti-LAMTOR1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309727EA 636 EUR
ORIGTA309727
Anti-LAMTOR1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: LAMTOR1 (late endosomal/lysosomal adaptor, MAPK and MTOR activator 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR