Anti-KDM6B Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309713EA 697 EUR
ORIGTA309713
Anti-KDM6B Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: KDM6B (lysine (K)-specific demethylase 6B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR