Anti-KDM4C Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309711EA 723 EUR
ORIGTA309711
Anti-KDM4C Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: KDM4C (lysine (K)-specific demethylase 4C)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR