Anti-HIP1 Mouse Monoclonal Antibody [clone: 4B10]

Lieferant: OriGene
ORIGTA309695EA 636 EUR
ORIGTA309695
Anti-HIP1 Mouse Monoclonal Antibody [clone: 4B10]
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: HIP1 (huntingtin interacting protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR