Anti-GLDN Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309672EA 636 EUR
ORIGTA309672
Anti-GLDN Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: GLDN (gliomedin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR