Anti-EGLN3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309638EA 723 EUR
ORIGTA309638
Anti-EGLN3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: EGLN3 (egl-9 family hypoxia-inducible factor 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR