Anti-CDX1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309604EA 577 EUR
ORIGTA309604
Anti-CDX1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: CDX1 (caudal type homeobox 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR