Anti-BHLHE40 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309587EA 761 EUR
ORIGTA309587
Anti-BHLHE40 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: BHLHE40 (basic helix-loop-helix family, member e40)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR