Anti-ARID4B Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309575EA 544 EUR
ORIGTA309575
Anti-ARID4B Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: ARID4B (AT rich interactive domain 4B (RBP1-like))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR