Anti-ABCA1 Mouse Monoclonal Antibody [clone: HJ1]

Lieferant: OriGene
ORIGTA309556EA 668 EUR
ORIGTA309556
Anti-ABCA1 Mouse Monoclonal Antibody [clone: HJ1]
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: ABCA1 (ATP-binding cassette, sub-family A (ABC1), member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR