Anti-AANAT Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309552EA 668 EUR
ORIGTA309552
Anti-AANAT Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: AANAT (aralkylamine N-acetyltransferase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR