Anti-CHIT1 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA309549EA 536 EUR
ORIGTA309549
Anti-CHIT1 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: CHIT1 (chitinase 1 (chitotriosidase))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR