Anti-ZNF652 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA309538EA 536 EUR
ORIGTA309538
Anti-ZNF652 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.01-0.03?g/ml

Type: Primary
Antigen: ZNF652 (zinc finger protein 652)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR