Anti-NPPA Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA309531EA 536 EUR
ORIGTA309531
Anti-NPPA Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.01-0.03?g/ml; Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: NPPA (natriuretic peptide A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR