Anti-PRODH Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA309525EA 536 EUR
ORIGTA309525
Anti-PRODH Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.3-1?g/ml

Type: Primary
Antigen: PRODH (proline dehydrogenase (oxidase) 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR