Anti-DOC2A Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA309524EA 539 EUR
ORIGTA309524
Anti-DOC2A Goat Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: DOC2A (double C2-like domains, alpha)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR