Anti-WFDC1 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA309509EA 536 EUR
ORIGTA309509
Anti-WFDC1 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.3-1?g/ml

Type: Primary
Antigen: WFDC1 (WAP four-disulfide core domain 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR