Anti-KCNT2 Mouse Monoclonal Antibody [clone: S11-33]
Lieferant: OriGene
ORIGTA309478EA 678 EUR
ORIGTA309478
Anti-KCNT2 Mouse Monoclonal Antibody [clone: S11-33]
Antikörper
Recommended Dilutions:
Type: Primary
Antigen: KCNT2 (potassium channel, subfamily T, member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat