Anti-DNAJA2 Mouse Monoclonal Antibody [clone: 1G10.H8]

Lieferant: OriGene
ORIGTA309369EA 648 EUR
ORIGTA309369
Anti-DNAJA2 Mouse Monoclonal Antibody [clone: 1G10.H8]
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: DNAJA2 (DnaJ (Hsp40) homolog, subfamily A, member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Yeast
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR