Anti-DUX4 Mouse Antibody [clone: P2B1]

Lieferant: OriGene
ORIGTA309324EA 678 EUR
ORIGTA309324
Anti-DUX4 Mouse Antibody [clone: P2B1]
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: DUX4 (double homeobox 4)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR