Anti-CACNA1D Mouse Monoclonal Antibody [clone: S48A-9]
Lieferant: OriGene
ORIGTA309307EA 678 EUR
ORIGTA309307
Anti-CACNA1D Mouse Monoclonal Antibody [clone: S48A-9]
Antikörper
Recommended Dilutions: Western Blot: 1-10?g/ml; Immunochemistry: 0.1-1.0?g/ml; Immunofluorescence: 1.0-10?g/ml
Type: Primary
Antigen: CACNA1D (calcium channel, voltage-dependent, L type, alpha 1D subunit)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat