Anti-UCHL1 Mouse Antibody [clone: BH7]

Lieferant: OriGene
ORIGTA309271EA 645 EUR
ORIGTA309271
Anti-UCHL1 Mouse Antibody [clone: BH7]
Antikörper
Recommended Dilutions: Western Blot: 1:5000; Immunofluorescence: 1:500

Type: Primary
Antigen: UCHL1 (ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase))
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR