Anti-THRA Mouse Antibody [clone: 1330]

Lieferant: OriGene
ORIGTA309256EA 645 EUR
ORIGTA309256
Anti-THRA Mouse Antibody [clone: 1330]
Antikörper
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: THRA (thyroid hormone receptor, alpha)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR