Anti-VAMP1/2 Mouse Antibody [clone: SP10]

Lieferant: OriGene
ORIGTA309244EA 645 EUR
ORIGTA309244
Anti-VAMP1/2 Mouse Antibody [clone: SP10]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:100

Type: Primary
Antigen: VAMP1/2
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR