Anti-SYN1 Rabbit Antibody

Lieferant: OriGene
ORIGTA309239EA 567 EUR
ORIGTA309239
Anti-SYN1 Rabbit Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: SYN1 (synapsin I)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR