Anti-RHO Mouse Antibody [clone: 1D4]

Lieferant: OriGene
ORIGTA309233EA 645 EUR
ORIGTA309233
Anti-RHO Mouse Antibody [clone: 1D4]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:100

Type: Primary
Antigen: RHO (rhodopsin)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR