Anti-RAF1 Rabbit Antibody

Lieferant: OriGene
ORIGTA309228EA 645 EUR
ORIGTA309228
Anti-RAF1 Rabbit Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: RAF1 (v-raf-1 murine leukemia viral oncogene homolog 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR