Anti-RAB3A Rabbit Antibody

Lieferant: OriGene
ORIGTA309224EA 622 EUR
ORIGTA309224
Anti-RAB3A Rabbit Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: RAB3A (RAB3A, member RAS oncogene family)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR