Anti-OLIG1 Rabbit Antibody

Lieferant: OriGene
ORIGTA309204EA 645 EUR
ORIGTA309204
Anti-OLIG1 Rabbit Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:3000; Immunochemistry: 1:1000; Immunoprecipitation: 1:50

Type: Primary
Antigen: OLIG1 (oligodendrocyte transcription factor 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR