Anti-GRIN2B Rabbit Antibody

Lieferant: OriGene
ORIGTA309196EA 645 EUR
ORIGTA309196
Anti-GRIN2B Rabbit Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: GRIN2B (glutamate receptor, ionotropic, N-methyl D-aspartate 2B)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR