Anti-GRIN2A Rabbit Antibody

Lieferant: OriGene
ORIGTA309190EA 645 EUR
ORIGTA309190
Anti-GRIN2A Rabbit Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: GRIN2A (glutamate receptor, ionotropic, N-methyl D-aspartate 2A)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR