Anti-NEFH Chicken Antibody

Lieferant: OriGene
ORIGTA309177EA 567 EUR
ORIGTA309177
Anti-NEFH Chicken Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:50000; Immunofluorescence: 1:25000

Type: Primary
Antigen: NEFH (neurofilament, heavy polypeptide)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR