Anti-MBP Chicken Antibody

Lieferant: OriGene
ORIGTA309175EA 548 EUR
ORIGTA309175
Anti-MBP Chicken Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:5000; Immunofluorescence: 1:500

Type: Primary
Antigen: MBP (myelin basic protein)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR