Anti-GRM7 Rabbit Antibody

Lieferant: OriGene
ORIGTA309173EA 645 EUR
ORIGTA309173
Anti-GRM7 Rabbit Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: GRM7 (glutamate receptor, metabotropic 7)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR