Anti-MAP2K1 Rabbit Antibody

Lieferant: OriGene
ORIGTA309168EA 645 EUR
ORIGTA309168
Anti-MAP2K1 Rabbit Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: MAP2K1 (mitogen-activated protein kinase kinase 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR