Anti-SLC6A13 Rabbit Antibody

Lieferant: OriGene
ORIGTA309146EA 645 EUR
ORIGTA309146
Anti-SLC6A13 Rabbit Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:100-1:200

Type: Primary
Antigen: SLC6A13 (solute carrier family 6 (neurotransmitter transporter), member 13)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR