Anti-GABRA6 Rabbit Antibody

Lieferant: OriGene
ORIGTA309131EA 567 EUR
ORIGTA309131
Anti-GABRA6 Rabbit Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: GABRA6 (gamma-aminobutyric acid (GABA) A receptor, alpha 6)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR