Anti-CLVS1/2 Rabbit Antibody

Lieferant: OriGene
ORIGTA309094EA 658 EUR
ORIGTA309094
Anti-CLVS1/2 Rabbit Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000 Immunochemistry: 1:50

Type: Primary
Antigen: CLVS1/2
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR