Anti-CALB2 Sheep Antibody

Lieferant: OriGene
ORIGTA309086EA 658 EUR
ORIGTA309086
Anti-CALB2 Sheep Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:500; Immunofluorescence: 1:500

Type: Primary
Antigen: CALB2 (calbindin 2)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR