Anti-ACTA1 Mouse Antibody [clone: C4]

Lieferant: OriGene
ORIGTA309077EA 567 EUR
ORIGTA309077
Anti-ACTA1 Mouse Antibody [clone: C4]
Antikörper
Recommended Dilutions: Western Blot:1:5000; Immunofluorescence: 1:500; Immunochemistry: 1:500

Type: Primary
Antigen: ACTA1 (actin, alpha 1, skeletal muscle)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR