Anti-INA Mouse Antibody [clone: ID2]

Lieferant: OriGene
ORIGTA309074EA 645 EUR
ORIGTA309074
Anti-INA Mouse Antibody [clone: ID2]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:250; Immunochemistry: 1:1000

Type: Primary
Antigen: INA (internexin neuronal intermediate filament protein, alpha)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR