Anti-YIPF4 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309041EA 667 EUR
ORIGTA309041
Anti-YIPF4 Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Immunochemistry1:100-1:1000; Western Blot:1:500-1:3000

Type: Primary
Antigen: YIPF4 (Yip1 domain family, member 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR